2-Amino-N,N-dimethylbenzolsulfonamid

Lieferant: ACTIVATE SCIENTIFIC

AS30770G1 AS30770G5
ACTIAS30770G1EA 0 CHF
ACTIAS30770G1 ACTIAS30770G5
2-Amino-N,N-dimethylbenzolsulfonamid
2-Amino-N,N-dimethylbenzolsulfonamid
Formel: C₈H₁₂N₂O₂S
MDL Number: MFCD04035363
CAS-Nummer: 54468-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR