Methyl-3-fluor-2-nitrobenzoat

Lieferant: ACTIVATE SCIENTIFIC

AS34455G1 AS34455G5
ACTIAS34455G1EA 98.92 CHF
ACTIAS34455G1 ACTIAS34455G5
Methyl-3-fluor-2-nitrobenzoat
Methyl-3-fluor-2-nitrobenzoat
Formel: C₈H₆FNO₄
MDL Number: MFCD11520393
CAS-Nummer: 1214353-57-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR