Methyl-2-fluor-4-nitrobenzoat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 2-fluoro-4-nitrobenzoate

AS34446G1 AS34446G10 AS34446G5
ACTIAS34446G1EA 0 CHF
ACTIAS34446G1 ACTIAS34446G10 ACTIAS34446G5
Methyl-2-fluor-4-nitrobenzoat
Methyl-2-fluor-4-nitrobenzoat
Formel: C₈H₆FNO₄
MW: 199.1400 g/mol
MDL Number: MFCD08444028
CAS-Nummer: 392-09-6
EINECS: 206-873-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR