Methyl-2-fluor-3-nitrobenzoat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 2-fluoro-3-nitrobenzoate

AS31527G1 AS31527G5
ACTIAS31527G1EA 0 CHF
ACTIAS31527G1 ACTIAS31527G5
Methyl-2-fluor-3-nitrobenzoat
Methyl-2-fluor-3-nitrobenzoat
Formel: C₈H₆FNO₄
MW: 199.1400 g/mol
MDL Number: MFCD10566813
CAS-Nummer: 946126-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR