Methyl-2,4-difluor-5-nitrobenzoat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 2,4-difluoro-5-nitrobenzoate

AS33972G1 AS33972G5
ACTIAS33972G1EA 0 CHF
ACTIAS33972G1 ACTIAS33972G5
Methyl-2,4-difluor-5-nitrobenzoat
Methyl-2,4-difluor-5-nitrobenzoat
Formel: C₈H₅F₂NO₄
MW: 217.1264 g/mol
MDL Number: MFCD07779369
CAS-Nummer: 125568-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR