4-Chlor-2-hydroxybenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS36308G1 AS36308G5
ACTIAS36308G1EA 0 CHF
ACTIAS36308G1 ACTIAS36308G5
4-Chlor-2-hydroxybenzonitril
4-Chlor-2-hydroxybenzonitril
Formel: C₇H₄ClNO
MW: 153.5700 g/mol
MDL Number: MFCD00234255
CAS-Nummer: 30818-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR