3-Chlor-5-hydroxybenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS32385G1 AS32385G5
ACTIAS32385G1EA 0 CHF
ACTIAS32385G1 ACTIAS32385G5
3-Chlor-5-hydroxybenzonitril
3-Chlor-5-hydroxybenzonitril
Formel: C₇H₄ClNO
MW: 153.5658 g/mol
MDL Number: MFCD11226540
CAS-Nummer: 473923-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR