2-Brom-5-hydroxybenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS30792G1 AS30792G5
ACTIAS30792G1EA 0 CHF
ACTIAS30792G1 ACTIAS30792G5
2-Brom-5-hydroxybenzonitril
2-Brom-5-hydroxybenzonitril
Formel: C₇H₄BrNO
MW: 198.0200 g/mol
MDL Number: MFCD08061922
CAS-Nummer: 189680-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR