2-Brom-4-methoxybenzonitril
Lieferant: ACTIVATE SCIENTIFIC
AS34081G1
AS34081G5
ACTIAS34081G1EA
0
CHF
ACTIAS34081G1
ACTIAS34081G5
2-Brom-4-methoxybenzonitril
2-Brom-4-methoxybenzonitril
Formel:
C₈H₆BrNO MW: 212,0433 g/mol |
MDL Number:
MFCD09056764 CAS-Nummer: 140860-51-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...