3-Chlor-6-phenylpyridazin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Phenyl-6-chloropyridazine

AS30949G5 AS30949G1
ACTIAS30949G5EA 0 CHF
ACTIAS30949G5 ACTIAS30949G1
3-Chlor-6-phenylpyridazin
3-Chloro-6-phenylpyridazine
Formel: C₁₀H₇ClN₂
MW: 190,6300 g/mol
MDL Number: MFCD00075253
CAS-Nummer: 20375-65-9
EINECS: 243-772-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR