8-(Brommethyl)chinolin

Lieferant: ACTIVATE SCIENTIFIC

AS1356G1 AS1356G5
ACTIAS1356G1EA 0 CHF
ACTIAS1356G1 ACTIAS1356G5
8-(Brommethyl)chinolin
8-(Bromomethyl)quinoline
Formel: C₁₀H₈BrN
MW: 222,0800 g/mol
MDL Number: MFCD00047621
CAS-Nummer: 7496-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR