5-Fluor-3-pyridinboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Fluoropyridine-3-boronic acid, (5-Fluoropyridin-3-yl)boronic acid

AS36392G1 AS36392G5
ACTIAS36392G1EA 0 CHF
ACTIAS36392G1 ACTIAS36392G5
5-Fluor-3-pyridinboronsäure
5-Fluoro-3-pyridineboronic acid
Formel: C₅H₅BFNO₂
MW: 140.91 g/mol
MDL Number: MFCD07368243
CAS-Nummer: 872041-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR