2,6-Difluor-3-pyridinboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2,6-Difluoropyridine-3-boronic acid

AS50719G1 AS50719G25 AS50719G5
ACTIAS50719G1EA 0 CHF
ACTIAS50719G1 ACTIAS50719G25 ACTIAS50719G5
2,6-Difluor-3-pyridinboronsäure
2,6-Difluoro-3-pyridineboronic acid
Formel: C₅H₄BF₂NO₂
MW: 158.90 g/mol
MDL Number: MFCD03788245
CAS-Nummer: 136466-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR