5-Cyan-2-methylanilin

Lieferant: ACTIVATE SCIENTIFIC

AS32428G10 AS32428G5
ACTIAS32428G10EA 0 CHF
ACTIAS32428G10 ACTIAS32428G5
5-Cyan-2-methylanilin
5-Cyan-2-methylanilin
Formel: C₈H₈N₂
MDL Number: MFCD04039781
CAS-Nummer: 60710-80-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR