2-Cyan-6-methylanilin

Lieferant: ACTIVATE SCIENTIFIC

AS36549G1 AS36549G5
ACTIAS36549G1EA 0 CHF
ACTIAS36549G1 ACTIAS36549G5
2-Cyan-6-methylanilin
2-Cyan-6-methylanilin
Formel: C₈H₈N₂
MW: 132,1625 g/mol
MDL Number: MFCD09880087
CAS-Nummer: 69797-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR