5-Amino-2-brombenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS33444G1 AS33444G5
ACTIAS33444G1EA 0 CHF
ACTIAS33444G1 ACTIAS33444G5
5-Amino-2-brombenzonitril
5-Amino-2-brombenzonitril
Formel: C₇H₅BrN₂
MW: 197,0320 g/mol
MDL Number: MFCD00234259
CAS-Nummer: 72115-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR