3-Amino-5-brombenzonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Amino-3-bromobenzonitrile

AS31303G1 AS31303G5
ACTIAS31303G1EA 0 CHF
ACTIAS31303G1 ACTIAS31303G5
3-Amino-5-brombenzonitril
3-Amino-5-brombenzonitril
Formel: C₇H₅BrN₂
MW: 197.0300 g/mol
MDL Number: MFCD11109874
CAS-Nummer: 49674-16-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR