2-Amino-6-brombenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS21296G1 AS21296G25 AS21296G5
ACTIAS21296G1EA 0 CHF
ACTIAS21296G1 ACTIAS21296G25 ACTIAS21296G5
2-Amino-6-brombenzonitril
2-Amino-6-brombenzonitril
Formel: C₇H₅BrN₂
MDL Number: MFCD09834769
CAS-Nummer: 77326-62-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR