5-Cyan-2-nitroanilin

Lieferant: ACTIVATE SCIENTIFIC

AS34354G1 AS34354G5
ACTIAS34354G1EA 0 CHF
ACTIAS34354G1 ACTIAS34354G5
5-Cyan-2-nitroanilin
5-Cyan-2-nitroanilin
Formel: C₇H₅N₃O₂
MW: 163,1335 g/mol
MDL Number: MFCD18389919
CAS-Nummer: 99512-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR