2-Cyan-5-nitroanilin

Lieferant: ACTIVATE SCIENTIFIC

AS30150G1 AS30150G5
ACTIAS30150G1EA 0 CHF
ACTIAS30150G1 ACTIAS30150G5
2-Cyan-5-nitroanilin
2-Cyan-5-nitroanilin
Formel: C₇H₅N₃O₂
MW: 163,1300 g/mol
MDL Number: MFCD04117876
CAS-Nummer: 87376-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR