3-Hydroxy-4-methylbenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS34426G1 AS34426G5
ACTIAS34426G1EA 0 CHF
ACTIAS34426G1 ACTIAS34426G5
3-Hydroxy-4-methylbenzonitril
3-Hydroxy-4-methylbenzonitril
Formel: C₈H₇NO
MW: 133,1500 g/mol
MDL Number: MFCD08361769
CAS-Nummer: 3816-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR