2-Hydroxy-6-methylbenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS32463G1 AS32463G5
ACTIAS32463G1EA 0 CHF
ACTIAS32463G1 ACTIAS32463G5
2-Hydroxy-6-methylbenzonitril
2-Hydroxy-6-methylbenzonitril
Formel: C₈H₇NO
MDL Number: MFCD02261940
CAS-Nummer: 73289-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR