2-Hydroxy-4-methylbenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS33729G1 AS33729G5
ACTIAS33729G1EA 0 CHF
ACTIAS33729G1 ACTIAS33729G5
2-Hydroxy-4-methylbenzonitril
2-Hydroxy-4-methylbenzonitril
Formel: C₈H₇NO
MW: 133,1500 g/mol
MDL Number: MFCD02261941
CAS-Nummer: 18495-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR