3-Brom-5-(trifluormethyl)phenol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Bromo-5-hydroxybenzotrifluoride

AS36316G1 AS36316G5
ACTIAS36316G1EA 0 CHF
ACTIAS36316G1 ACTIAS36316G5
3-Brom-5-(trifluormethyl)phenol
3-Brom-5-(trifluormethyl)phenol
Formel: C₇H₄BrF₃O
MW: 241.0100 g/mol
MDL Number: MFCD11041083
CAS-Nummer: 1025718-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR