2-Brom-5-(trifluormethyl)phenol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Bromo-5-trifluoromethylphenol

AS31279G1 AS31279G5
ACTIAS31279G1EA 20 CHF
ACTIAS31279G1 ACTIAS31279G5
2-Brom-5-(trifluormethyl)phenol
2-Brom-5-(trifluormethyl)phenol
Formel: C₇H₄BrF₃O
MW: 241,01 g/mol
MDL Number: MFCD09261190
CAS-Nummer: 402-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR