6-Fluor-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS1905G1 AS1905G5
ACTIAS1905G1EA 0 CHF
ACTIAS1905G1 ACTIAS1905G5
6-Fluor-1H-indazol
6-Fluoro-1H-indazole
Formel: C₇H₅FN₂
MW: 136,1264 g/mol
MDL Number: MFCD07371562
CAS-Nummer: 348-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR