3-Methyl-5-isoxazolcarbonsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Methylisoxazole-5-carboxylic acid

AS31836G1 AS31836G5
ACTIAS31836G1EA 0 CHF
ACTIAS31836G1 ACTIAS31836G5
3-Methyl-5-isoxazolcarbonsäure
3-Methyl-5-isoxazolecarboxylic acid
Formel: C₅H₅NO₃
MW: 127,10 g/mol
MDL Number: MFCD00464222
CAS-Nummer: 4857-42-5
EINECS: 225-454-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR