3-Methyl-4-isoxazolcarbonsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Methylisoxazole-4-carboxylic acid

AS33721G1 AS33721G5
ACTIAS33721G1EA 0 CHF
ACTIAS33721G1 ACTIAS33721G5
3-Methyl-4-isoxazolcarbonsäure
3-Methyl-4-isoxazolecarboxylic acid
Formel: C₅H₅NO₃
MW: 127,10 g/mol
MDL Number: MFCD01318161
CAS-Nummer: 17153-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR