4-Isoxazolcarbonsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Isoxazole-4-carboxylic acid

AS31393G1 AS31393G5
ACTIAS31393G1EA 0 CHF
ACTIAS31393G1 ACTIAS31393G5
4-Isoxazolcarbonsäure
4-Isoxazolecarboxylic acid
Formel: C₄H₃NO₃
MW: 113,0715 g/mol
MDL Number: MFCD06738806
CAS-Nummer: 6436-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR