3-Brom-5-nitro-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Bromo-5-nitroindazole

AS23917G100 AS23917G25 AS23917G5
ACTIAS23917G100EA 248 CHF
ACTIAS23917G100 ACTIAS23917G25 ACTIAS23917G5
3-Brom-5-nitro-1H-indazol
3-Bromo-5-nitro-1H-indazole
Formel: C₇H₄BrN₃O₂
MW: 242,03 g/mol
MDL Number: MFCD07367964
CAS-Nummer: 67400-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR