6-Bromnicotinonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Bromo-5-cyanopyridine

AS4049G1 AS4049G25 AS4049G5
ACTIAS4049G1EA 18 CHF
ACTIAS4049G1 ACTIAS4049G25 ACTIAS4049G5
6-Bromnicotinonitril
6-Bromnicotinonitril
Formel: C₆H₃BrN₂
MW: 183,01 g/mol
MDL Number: MFCD00234141
CAS-Nummer: 139585-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR