2-Bromnicotinonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Bromo-3-cyanopyridine

AS30541G1 AS30541G5
ACTIAS30541G1EA 0 CHF
ACTIAS30541G1 ACTIAS30541G5
2-Bromnicotinonitril
2-Bromnicotinonitril
Formel: C₆H₃BrN₂
MW: 183,01 g/mol
MDL Number: MFCD00234140
CAS-Nummer: 20577-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR