7-Brom-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS50934G1 AS50934G5
ACTIAS50934G1EA 0 CHF
ACTIAS50934G1 ACTIAS50934G5
7-Brom-1H-indazol
7-Bromo-1H-indazole
Formel: C₇H₅BrN₂
MW: 197,0300 g/mol
MDL Number: MFCD07371564
CAS-Nummer: 53857-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR