3-Brom-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS8065G1 AS8065G5
ACTIAS8065G1EA 0 CHF
ACTIAS8065G1 ACTIAS8065G5
3-Brom-1H-indazol
3-Bromo-1H-indazole
Formel: C₇H₅BrN₂
MDL Number: MFCD00159926
CAS-Nummer: 40598-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR