3,5-Dibrom-1H-indazol

Lieferant: ACTIVATE SCIENTIFIC

AS30778G1 AS30778G5
ACTIAS30778G1EA 0 CHF
ACTIAS30778G1 ACTIAS30778G5
3,5-Dibrom-1H-indazol
3,5-Dibromo-1H-indazole
Formel: C₇H₄Br₂N₂
MDL Number: MFCD00159919
CAS-Nummer: 40598-76-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR