2,6-Dibromisonicotinsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2,6-Dibromopyridine-4-carboxylic acid

AS31199G1 AS31199G5
ACTIAS31199G1EA 0 CHF
ACTIAS31199G1 ACTIAS31199G5
2,6-Dibromisonicotinsäure
2,6-Dibromisonicotinsäure
Formel: C₆H₃Br₂NO₂
MW: 280,9000 g/mol
MDL Number: MFCD09743987
CAS-Nummer: 2016-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR