2,4-Difluor-3-methylbenzonitril
Lieferant: ACTIVATE SCIENTIFIC
AS35446G1
AS35446G5
ACTIAS35446G1EA
0
CHF
ACTIAS35446G1
ACTIAS35446G5
2,4-Difluor-3-methylbenzonitril
2,4-Difluor-3-methylbenzonitril
Formel:
C₈H₅F₂N MW: 153,1288 g/mol |
MDL Number:
MFCD07777155 CAS-Nummer: 847502-87-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...