4-Chlor-2-methylbenzonitril
Lieferant: ACTIVATE SCIENTIFIC
AS24961G100
AS24961G25
AS24961G5
ACTIAS24961G100EA
218
CHF
ACTIAS24961G100
ACTIAS24961G25
ACTIAS24961G5
4-Chlor-2-methylbenzonitril
4-Chlor-2-methylbenzonitril
Formel:
C₈H₆ClN MW: 151,60 g/mol |
MDL Number:
MFCD00045603 CAS-Nummer: 50712-68-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...