4-Chlor-3-methylbenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS32624G1 AS32624G5
ACTIAS32624G1EA 0 CHF
ACTIAS32624G1 ACTIAS32624G5
4-Chlor-3-methylbenzonitril
4-Chlor-3-methylbenzonitril
Formel: C₈H₆ClN
MW: 151.5900 g/mol
MDL Number: MFCD03452793
CAS-Nummer: 4387-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR