2-Chlor-4-methylbenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS33744G1 AS33744G5
ACTIAS33744G1EA 0 CHF
ACTIAS33744G1 ACTIAS33744G5
2-Chlor-4-methylbenzonitril
2-Chlor-4-methylbenzonitril
Formel: C₈H₆CIN
MW: 151,5900 g/mol
MDL Number: MFCD03094672
CAS-Nummer: 21423-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR