2-Iod-5-methoxybenzoesäure

Lieferant: ACTIVATE SCIENTIFIC

AS35742G1 AS35742G10 AS35742G5
ACTIAS35742G1EA 0 CHF
ACTIAS35742G1 ACTIAS35742G10 ACTIAS35742G5
2-Iod-5-methoxybenzoesäure
2-Iodo-5-methoxybenzoic acid
Formel: C₈H₇IO₃
MW: 278,0400 g/mol
MDL Number: MFCD00204590
CAS-Nummer: 54413-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR