2-Fluor-4-hydroxybenzoesäure

Lieferant: ACTIVATE SCIENTIFIC

AS25267G25 AS25267G5
ACTIAS25267G25EA 130 CHF
ACTIAS25267G25 ACTIAS25267G5
2-Fluor-4-hydroxybenzoesäure
2-Fluoro-4-hydroxybenzoic acid
Formel: C₇H₅FO₃
MW: 156,1112 g/mol
MDL Number: MFCD01310985
CAS-Nummer: 65145-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR