5-Fluor-m-toluidin

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS32397G1EA 0 CHF
ACTIAS32397G1 ACTIAS32397G5
5-Fluor-m-toluidin
5-Fluor-m-toluidin
Formel: C₇H₈FN
MW: 125,14 g/mol
MDL Number: MFCD00077538
CAS-Nummer: 52215-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR