2-(Difluormethoxy)phenylessigsäure

Lieferant: ACTIVATE SCIENTIFIC

AS34369G1 AS34369G5
ACTIAS34369G1EA 0 CHF
ACTIAS34369G1 ACTIAS34369G5
2-(Difluormethoxy)phenylessigsäure
2-(Difluormethoxy)phenylessigsäure
Formel: C₉H₈F₂O₃
MDL Number: MFCD00236237
CAS-Nummer: 86867-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR