(3,5-Dichlorphenyl)essigsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-(3,5-Dichlorophenyl)acetic acid

AS32175G1 AS32175G5
ACTIAS32175G1EA 20 CHF
ACTIAS32175G1 ACTIAS32175G5
(3,5-Dichlorphenyl)essigsäure
(3,5-Dichlorphenyl)essigsäure
Formel: C₈H₆Cl₂O₂
MW: 205.03 g/mol
MDL Number: MFCD01764671
CAS-Nummer: 51719-65-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR