3,5-Dichlorpyridazin

Lieferant: ACTIVATE SCIENTIFIC

AS33726G1 AS33726G5
ACTIAS33726G1EA 0 CHF
ACTIAS33726G1 ACTIAS33726G5
3,5-Dichlorpyridazin
3,5-Dichloropyridazine
Formel: C₄H₂Cl₂N₂
MDL Number: MFCD10698048
CAS-Nummer: 1837-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR