3,4,6-Trichlorpyridazin

Lieferant: ACTIVATE SCIENTIFIC

AS32111G1 AS32111G5
ACTIAS32111G1EA 0 CHF
ACTIAS32111G1 ACTIAS32111G5
3,4,6-Trichlorpyridazin
3,4,6-Trichloropyridazine
Formel: C₄HCl₃N₂
MW: 183.4231 g/mol
MDL Number: MFCD00834957
CAS-Nummer: 6082-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR