3-Amino-6-brom-4-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS33666G1 AS33666G5
ACTIAS33666G1EA 0 CHF
ACTIAS33666G1 ACTIAS33666G5
3-Amino-6-brom-4-methylpyridin
3-Amino-6-bromo-4-methylpyridine
Formel: C₆H₇BrN₂
MDL Number: MFCD08277280
CAS-Nummer: 156118-16-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR