3-Amino-6-brom-2-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Amino-2-bromo-6-methylpyridine, 6-Bromo-2-methylpyridin-3-amine

AS7235G1 AS7235G5
ACTIAS7235G1EA 0 CHF
ACTIAS7235G1 ACTIAS7235G5
3-Amino-6-brom-2-methylpyridin
3-Amino-6-bromo-2-methylpyridine
Formel: C₆H₇BrN₂
MW: 187,04 g/mol
MDL Number: MFCD03095086
CAS-Nummer: 126325-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR