3-Amino-2-brom-5-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Amino-2-bromo-5-picoline, 5-Amino-6-bromo-3-picoline, 2-Bromo-5-methyl-3-pyridinamine

AS33539G1 AS33539G10 AS33539G5
ACTIAS33539G1EA 0 CHF
ACTIAS33539G1 ACTIAS33539G10 ACTIAS33539G5
3-Amino-2-brom-5-methylpyridin
3-Amino-2-bromo-5-methylpyridine
Formel: C₆H₇BrN₂
MW: 187.04 g/mol
MDL Number: MFCD08277281
CAS-Nummer: 34552-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR